Anti-PPP1R7 Mouse Monoclonal Antibody [clone: 3E7]

Proveedor: OriGene
CF500605
ORIGCF500605EA 786 EUR
ORIGCF500605
Anti-PPP1R7 Mouse Monoclonal Antibody [clone: 3E7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PPP1R7
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR