Anti-CDCP1 Mouse Monoclonal Antibody [clone: 3B5]

Proveedor: OriGene
CF502230
ORIGCF502230EA 764 EUR
ORIGCF502230
Anti-CDCP1 Mouse Monoclonal Antibody [clone: 3B5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150

Type: Primary
Antigen: CDCP1
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR