Anti-ACAT2 Mouse Monoclonal Antibody [clone: 3E2]

Proveedor: OriGene
CF501222
ORIGCF501222EA 764 EUR
ORIGCF501222
Anti-ACAT2 Mouse Monoclonal Antibody [clone: 3E2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACAT2
Clonality: Monoclonal
Clone: 3E2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR