Anti-ACAT2 Mouse Monoclonal Antibody [clone: 5F7]

Proveedor: OriGene
CF501219
ORIGCF501219EA 764 EUR
ORIGCF501219
Anti-ACAT2 Mouse Monoclonal Antibody [clone: 5F7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACAT2
Clonality: Monoclonal
Clone: 5F7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR