Anti-PDE4A Mouse Monoclonal Antibody [clone: 5C6]

Proveedor: OriGene
CF501206
ORIGCF501206EA 764 EUR
ORIGCF501206
Anti-PDE4A Mouse Monoclonal Antibody [clone: 5C6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE4A
Clonality: Monoclonal
Clone: 5C6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR