Anti-SATB1 Mouse Monoclonal Antibody [clone: 3E12]

Proveedor: OriGene
ORIGCF500578EA 764 EUR
ORIGCF500578
Anti-SATB1 Mouse Monoclonal Antibody [clone: 3E12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: SATB1
Clonality: Monoclonal
Clone: 3E12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR