Anti-IFT57 Mouse Monoclonal Antibody [clone: 5B6]

Proveedor: OriGene
CF500906
ORIGCF500906EA 764 EUR
ORIGCF500906
Anti-IFT57 Mouse Monoclonal Antibody [clone: 5B6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: IFT57
Clonality: Monoclonal
Clone: 5B6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR