Anti-PLEK Mouse Monoclonal Antibody [clone: 2F10]

Proveedor: OriGene
CF500901
ORIGCF500901EA 786 EUR
ORIGCF500901
Anti-PLEK Mouse Monoclonal Antibody [clone: 2F10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: PLEK
Clonality: Monoclonal
Clone: 2F10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR