Anti-MSI1 Mouse Monoclonal Antibody [clone: 1G8]

Proveedor: OriGene
CF502198
ORIGCF502198EA 764 EUR
ORIGCF502198
Anti-MSI1 Mouse Monoclonal Antibody [clone: 1G8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MSI1
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR