Anti-MAPK7 Mouse Monoclonal Antibody [clone: 4C1]

Proveedor: OriGene
CF502187
ORIGCF502187EA 764 EUR
ORIGCF502187
Anti-MAPK7 Mouse Monoclonal Antibody [clone: 4C1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPK7
Clonality: Monoclonal
Clone: 4C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR