Anti-NME4 Mouse Monoclonal Antibody [clone: 2B10]

Proveedor: OriGene
ORIGCF501177EA 764 EUR
ORIGCF501177
Anti-NME4 Mouse Monoclonal Antibody [clone: 2B10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NME4
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR