Anti-ACSBG1 Mouse Monoclonal Antibody [clone: 2B5]

Proveedor: OriGene
ORIGCF501175EA 764 EUR
ORIGCF501175
Anti-ACSBG1 Mouse Monoclonal Antibody [clone: 2B5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: ACSBG1
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR