Anti-PNMT Mouse Monoclonal Antibody [clone: 1E8]

Proveedor: OriGene
CF502849
ORIGCF502849EA 764 EUR
ORIGCF502849
Anti-PNMT Mouse Monoclonal Antibody [clone: 1E8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PNMT
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR