Anti-SULT1A1 Mouse Monoclonal Antibody [clone: 6C8]

Proveedor: OriGene
ORIGCF502159EA 764 EUR
ORIGCF502159
Anti-SULT1A1 Mouse Monoclonal Antibody [clone: 6C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT1A1
Clonality: Monoclonal
Clone: 6C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR