Anti-GSTT2 Mouse Monoclonal Antibody [clone: 8A10]

Proveedor: OriGene
CF501831
ORIGCF501831EA 786 EUR
ORIGCF501831
Anti-GSTT2 Mouse Monoclonal Antibody [clone: 8A10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GSTT2
Clonality: Monoclonal
Clone: 8A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR