Anti-TCP11L2 Mouse Monoclonal Antibody [clone: 1G3]

Proveedor: OriGene
CF501807
ORIGCF501807EA 786 EUR
ORIGCF501807
Anti-TCP11L2 Mouse Monoclonal Antibody [clone: 1G3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: TCP11L2
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR