Anti-BEST3 Mouse Monoclonal Antibody [clone: 3B11]

Proveedor: OriGene
CF501768
ORIGCF501768EA 786 EUR
ORIGCF501768
Anti-BEST3 Mouse Monoclonal Antibody [clone: 3B11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BEST3
Clonality: Monoclonal
Clone: 3B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR