Anti-IgG Antibody (HRP (Horseradish Peroxidase))

Proveedor: Boster
BSBTBA1060-0.5EA 0 EUR
BSBTBA1060-0.5
Anti-IgG Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR