Anti- IgG Antibody (Unconjugated)

Proveedor: Boster
BSBTBA1048-5EA 0 EUR
BSBTBA1048-5
Anti- IgG Antibody (Unconjugated)
Anticuerpos
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR