Anti-IgM Antibody (Biotin)

Proveedor: Boster
BSBTBA1004-0.25EA 0 EUR
BSBTBA1004-0.25
Anti-IgM Antibody (Biotin)
Anticuerpos
Type: Secondary
Antigen: IgM
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR