Anti-PGR Mouse Monoclonal Antibody [clone: 5F82]

Proveedor: United States Biological
USBIP9007-01A1EA 709 EUR
USBIP9007-01A1
Anti-PGR Mouse Monoclonal Antibody [clone: 5F82]
Anticuerpos
Recommended dilution: Immunochemistry: 1:5-1:10

Type: Primary
Antigen: PGR (progesterone receptor)
Clonality: monoclonal
Clone: 5F82
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR