Anti-FOXO3 Rabbit Polyclonal Antibody

Proveedor: United States Biological
USBI146679EA 526 EUR
USBI146679
Anti-FOXO3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: FOXO3 (forkhead box O3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR