Anti-PLRG1 Mouse monoclonal antibody
MEDMMM-0157-PEA 312 EUR
MEDMMM-0157-P
Anti-PLRG1 Mouse monoclonal antibody
Type: Primary
Antigen: PLRG1 (pleiotropic regulator 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now