Anti-IKBKG Mouse monoclonal antibody
MEDMMM-0150-PEA 284 EUR
MEDMMM-0150-P
Anti-IKBKG Mouse monoclonal antibody
Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Zebrafish