Anti-PPP3CA Rabbit Monoclonal Antibody

Proveedor: Medimabs
MEDMMM-0146-PEA 329 EUR
MEDMMM-0146-P
Anti-PPP3CA Rabbit Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: PPP3CA (protein phosphatase 3, catalytic subunit, alpha isozyme)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR