Anti-Smg-GDS Rabbit Polyclonal Antibody

Proveedor: Medimabs
MEDMMM-0140EA 243 EUR
MEDMMM-0140
Anti-Smg-GDS Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Smg-GDS
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR