Anti-VAT1 Rabbit Polyclonal Antibody

Proveedor: Medimabs
MEDMMM-0134EA 243 EUR
MEDMMM-0134
Anti-VAT1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: VAT1 (vesicle amine transport 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR