Anti-SceD Chicken Polyclonal Antibody

Proveedor: Medimabs
MEDMMM-0067EA 303 EUR
MEDMMM-0067
Anti-SceD Chicken Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SceD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR