Anti-CD160 Rat Monoclonal Antibody

Proveedor: Medimabs
MEDMMM-0062-PEA 268 EUR
MEDMMM-0062-P
Anti-CD160 Rat Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD160 (CD160 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR