Anti-H1P1 Rabbit Polyclonal Antibody

Proveedor: Medimabs
MEDMMM-0031EA 310 EUR
MEDMMM-0031
Anti-H1P1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: H1P1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR