Anti-GluR3B Mouse monoclonal antibody unconjugated

Proveedor: Medimabs
MEDMMM-0028-PEA 359 EUR
MEDMMM-0028-P
Anti-GluR3B Mouse monoclonal antibody unconjugated
Type: Primary
Antigen: GluR3B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR