Anti-SLC17A7 Mouse Monoclonal Antibody

Proveedor: Medimabs
MEDMMM-0016-PEA 418 EUR
MEDMMM-0016-P
Anti-SLC17A7 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: SLC17A7 (solute carrier family 17 (vesicular glutamate transporter), member 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR