Anti-HRP Mouse Monoclonal Antibody

Proveedor: Medimabs
MEDMMM-0012-PEA 384 EUR
MEDMMM-0012-P
Anti-HRP Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: HRP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR