Anti-Leu-enkehalin Mouse Monoclonal Antibody
Proveedor: Medimabs
MEDMMM-0006-PEA 425 EUR
MEDMMM-0006-P
Anti-Leu-enkehalin Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: Leu-enkehalin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat