Anti-HMCES Rat Monoclonal Antibody [clone: HMC-MA01]
Proveedor: United States Biological
USBI045667EA 751 EUR
USBI045667
Anti-HMCES Rat Monoclonal Antibody [clone: HMC-MA01]
Anticuerpos
Recommended dilution: Dot Blot: 1:5000
Type: Primary
Antigen: HMCES (5-hydroxymethylcytosine (hmC) binding, ES cell-specific)
Clonality: Monoclonal
Clone: HMC-MA01
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity:
Order Now