Anti-ARHGAP27 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Proveedor: United States Biological
USBI041692-APEA 852 EUR
USBI041692-AP
Anti-ARHGAP27 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: ARHGAP27 (Rho GTPase activating protein 27)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human