Anti-SH3BP1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Proveedor: United States Biological
USBI041689-APEA 852 EUR
USBI041689-AP
Anti-SH3BP1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SH3BP1 (SH3-domain binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human