Anti-SH3BP1 Rabbit Polyclonal Antibody

Proveedor: United States Biological
USBI041689EA 587 EUR
USBI041689
Anti-SH3BP1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SH3BP1 (SH3-domain binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR