Anti-SH3BGR Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Proveedor: United States Biological
USBI041686-PEEA 852 EUR
USBI041686-PE
Anti-SH3BGR Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SH3BGR (SH3 domain binding glutamate-rich protein)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR