Anti-SH2D2A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Proveedor: United States Biological
USBI041682-APCEA 852 EUR
USBI041682-APC
Anti-SH2D2A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SH2D2A (SH2 domain containing 2A)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human