Anti-SH2D2A Rabbit Polyclonal Antibody

Proveedor: United States Biological
USBI041682EA 587 EUR
USBI041682
Anti-SH2D2A Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SH2D2A (SH2 domain containing 2A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR