Anti-SERPINB9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Proveedor: United States Biological
USBI041583-FITCEA 852 EUR
USBI041583-FITC
Anti-SERPINB9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERPINB9 (serpin peptidase inhibitor, clade B (ovalbumin), member 9)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human