Anti-SERPINB12 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Proveedor: United States Biological
USBI041581-APCEA 852 EUR
USBI041581-APC
Anti-SERPINB12 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SERPINB12 (serpin peptidase inhibitor, clade B (ovalbumin), member 12)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR