Anti-SERPINB11 Rabbit Polyclonal Antibody (Biotin)
Proveedor: United States Biological
USBI041580-BIOTINEA 852 EUR
USBI041580-BIOTIN
Anti-SERPINB11 Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERPINB11 (serpin peptidase inhibitor, clade B (ovalbumin), member 11 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human