Anti-SERPINB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Proveedor: United States Biological
USBI041579-PEEA 852 EUR
USBI041579-PE
Anti-SERPINB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERPINB1 (serpin peptidase inhibitor, clade B (ovalbumin), member 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human