Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody (Biotin)
Proveedor: United States Biological
USBI041576-BIOTINEA 852 EUR
USBI041576-BIOTIN
Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50
Type: Primary
Antigen: Cortisol Binding Globulin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse