Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Proveedor: United States Biological
USBI041576-APCEA 852 EUR
USBI041576-APC
Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50
Do not freeze!
Type: Primary
Antigen: Cortisol Binding Globulin
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse