Anti-SERPINA5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Proveedor: United States Biological
USBI041575-APEA 852 EUR
USBI041575-AP
Anti-SERPINA5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERPINA5 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human