Anti-SERINC3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Proveedor: United States Biological
USBI041570-APCEA 852 EUR
USBI041570-APC
Anti-SERINC3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERINC3 (serine incorporator 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human