Anti-SERINC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: United States Biological
USBI041569-HRPEA 852 EUR
USBI041569-HRP
Anti-SERINC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERINC2 (serine incorporator 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human